Ethyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate

Supplier: Apollo Scientific
Warning

OR15189-10G OR15189-1G OR15189-5G
APOSOR15189-10GEA 814 GBP
APOSOR15189-10G APOSOR15189-1G APOSOR15189-5G
Ethyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate
Ethyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate
Formula: C₈H₁₂N₂O₂S
MW: 200,26 g/mol
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD08443952
CAS Number: 859521-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR