5-Amino-3-(3-chlorophenyl)isoxazole 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(3-Chlorophenyl)isoxazol-5-amine
OR15155-5G
OR15155-10G
APOSOR15155-5GEA
486
GBP
APOSOR15155-5G
APOSOR15155-10G
5-Amino-3-(3-chlorophenyl)isoxazole 95%
5-Amino-3-(3-chlorophenyl)isoxazole
Formula:
C₉H₇ClN₂O Melting Pt: 107…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD07383023 CAS Number: 86685-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...