7-Bromo-5-nitro-1H-indazole

Supplier: Apollo Scientific
Warning

OR15135-1G OR15135-500MG
APOSOR15135-1GEA 26.3 GBP
APOSOR15135-1G APOSOR15135-500MG
7-Bromo-5-nitro-1H-indazole
7-Bromo-5-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
Melting Pt: 255…257 °C
Storage Temperature: Ambient
MDL Number: MFCD03617603
CAS Number: 685109-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR