4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine

Supplier: Apollo Scientific
Warning

OR15121-1G OR15121-500MG
APOSOR15121-1GEA 148 GBP
APOSOR15121-1G APOSOR15121-500MG
4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine
4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine
Formula: C₁₅H₂₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02179169
CAS Number: 171049-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR