1,3-Oxazole-2-carbaldehyde 96%
Supplier: Apollo Scientific
|
Warning
|
OR14993-1G
OR14993-250MG
APOSOR14993-1GEA
0
GBP
APOSOR14993-1G
APOSOR14993-250MG
1,3-Oxazole-2-carbaldehyde 96%
1,3-Oxazole-2-carbaldehyde
Formula:
C₄H₃NO₂ Storage Temperature: Freezer |
MDL Number:
MFCD03412587 CAS Number: 65373-52-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...