2,3-Dihydro-1H-carbazol-4(9H)-one
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ondansetron Impurity I, 1,2,3,4-Tetrahydro-4-oxocarbazole, 1,2,3,4-Tetrahydrocarbazol-4-one
OR14906-100G
OR14906-25G
OR14906-5G
APOSOR14906-100GEA
97.1
GBP
APOSOR14906-100G
APOSOR14906-25G
APOSOR14906-5G
2,3-Dihydro-1H-carbazol-4(9H)-one
2,3-Dihydro-1H-carbazol-4(9H)-one
Formula:
C₁₂H₁₁NO MW: 185,2218 g/mol Melting Pt: 224…226 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00173749 CAS Number: 15128-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...