3-(tert-Butyl)-1,2,4-thiadiazol-5-amine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Amino-3-(tert-butyl)-1,2,4-thiadiazole
OR14854-1G
OR14854-250MG
APOSOR14854-1GEA
513
GBP
APOSOR14854-1G
APOSOR14854-250MG
3-(tert-Butyl)-1,2,4-thiadiazol-5-amine
3-(tert-Butyl)-1,2,4-thiadiazol-5-amine
Formula:
C₆H₁₁N₃S MW: 157,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09865006 CAS Number: 13383-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...