1-(2-Methoxyphenyl)-3-methylpiperazine
Supplier: Apollo Scientific
|
Warning
|
OR14834-1G
OR14834-250MG
APOSOR14834-1GEA
296
GBP
APOSOR14834-1G
APOSOR14834-250MG
1-(2-Methoxyphenyl)-3-methylpiperazine
1-(2-Methoxyphenyl)-3-methylpiperazine
Formula:
C₁₂H₁₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD06804057 CAS Number: 152943-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...