6-Nitro-2,4-xylidine 97%

Supplier: Apollo Scientific

Synonyms: 4,6-Dimethyl-2-nitroaniline, 1-Amino-2,4-dimethyl-6-nitrobenzene, (2,4-Dimethyl-6-nitrophenyl)amine, 2,4-Dimethyl-6-nitroaniline

OR14484-1G OR14484-5G
APOSOR14484-1GEA 49.9 GBP
APOSOR14484-1G APOSOR14484-5G
6-Nitro-2,4-xylidine 97%
6-Nitro-2,4-xylidine
Formula: C₈H₁₀N₂O₂
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007739
CAS Number: 1635-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR