2-(4-Methoxyphenyl)-4-quinolinecarboxylic acid

Supplier: Apollo Scientific
Warning

OR14384-10G OR14384-1G OR14384-5G
APOSOR14384-10GEA 0 GBP
APOSOR14384-10G APOSOR14384-1G APOSOR14384-5G
2-(4-Methoxyphenyl)-4-quinolinecarboxylic acid
2-(4-Methoxyphenyl)-4-quinolinecarboxylic acid
Formula: C₁₇H₁₃NO₃
MW: 279,3 g/mol
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00160581
CAS Number: 4364-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR