5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-2-hydroxy-3-methylbenzoic acid

OR14371-1G OR14371-5G
APOSOR14371-1GEA 73.5 GBP
APOSOR14371-1G APOSOR14371-5G
5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid 98%
5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid
Formula: C₈H₇BrO₃
MW: 231,05 g/mol
Melting Pt: 231…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00093269
CAS Number: 36194-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR