4-[(tert-Butoxycarbonylamino)methyl]benzoic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid, 4-[(Boc-amino)methyl]benzoic Acid, Boc-4-Amb-OH

OR14353-1G OR14353-25G OR14353-5G
APOSOR14353-1GEA 39.4 GBP
APOSOR14353-1G APOSOR14353-25G APOSOR14353-5G
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid 97%
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid
Formula: C₁₃H₁₇NO₄
MW: 251,28 g/mol
Melting Pt: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00228182
CAS Number: 33233-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR