2-Dimethylaminoquinoline-3-carbaldehyde 97%
Supplier: Apollo Scientific
|
Danger
|
OR14255-1G
OR14255-5G
APOSOR14255-1GEA
131
GBP
APOSOR14255-1G
APOSOR14255-5G
2-Dimethylaminoquinoline-3-carbaldehyde 97%
2-Dimethylaminoquinoline-3-carbaldehyde
Formula:
C₁₂H₁₂N₂O Melting Pt: 52…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD06010203 CAS Number: 728035-61-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...