3-Chloro-6-nitro-1H-indazole

Supplier: Apollo Scientific
Warning

OR14098-1G OR14098-250MG
APOSOR14098-1GEA 49.9 GBP
APOSOR14098-1G APOSOR14098-250MG
3-Chloro-6-nitro-1H-indazole
3-Chloro-6-nitro-1H-indazole
Formula: C₇H₄ClN₃O₂
MW: 197,5800 g/mol
Melting Pt: 203…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00010741
CAS Number: 50593-68-5
EINECS: 256-631-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR