N-[2-(Aminocarbonyl)phenyl]-4-chloro-3-nitrobenzenecarboxamide
Supplier: Apollo Scientific
Synonyms:
N-[2-(Aminocarbonyl)phenyl]-4-chloro-3-nitrobenzamide
OR14082-10G
OR14082-1G
OR14082-5G
APOSOR14082-10GEA
1520
GBP
APOSOR14082-10G
APOSOR14082-1G
APOSOR14082-5G
N-[2-(Aminocarbonyl)phenyl]-4-chloro-3-nitrobenzenecarboxamide
N-[2-(Aminocarbonyl)phenyl]-4-chloro-3-nitrobenzenecarboxamide
Formula:
C₁₄H₁₀ClN₃O₄ Melting Pt: 271…272 °C Storage Temperature: Ambient |
MDL Number:
MFCD00495266 CAS Number: 331240-40-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...