1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one 96%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one

OR14051-100G OR14051-25G OR14051-5G
APOSOR14051-100GEA 112 GBP
APOSOR14051-100G APOSOR14051-25G APOSOR14051-5G
1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one 96%
1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one
Formula: C₁₀H₁₂N₂O
MW: 176,22 g/mol
Melting Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00115180
CAS Number: 55314-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR