6-Methylpyridin-3-ol

Supplier: Apollo Scientific

OR13853-100G OR13853-25G
APOSOR13853-100GEA 68.4 GBP
APOSOR13853-100G APOSOR13853-25G
6-Methylpyridin-3-ol
6-Methylpyridin-3-ol
Formula: C₆H₇NO
Storage Temperature: Ambient
MDL Number: MFCD00006339
CAS Number: 1121-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR