Ethyl-3-bromo-1H-indole-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 3-bromoindole-2-carboxylate, Ethyl 3-Bromoindole-2-carboxylate, 3-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 3-bromo-1H-indole-2-carboxylate
OR13723-1G
OR13723-25G
OR13723-5G
APOSOR13723-1GEA
53.8
GBP
APOSOR13723-1G
APOSOR13723-25G
APOSOR13723-5G
Ethyl-3-bromo-1H-indole-2-carboxylate
Ethyl-3-bromo-1H-indole-2-carboxylate
Formula:
C₁₁H₁₀BrNO₂ MW: 268,11 g/mol Melting Pt: 149…151 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02071793 CAS Number: 91348-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...