3-Amino-4-chlorophenylboronic acid pinacol ester 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-4-chlorophenylboronic acid, pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

OR1336-1G OR1336-250MG OR1336-5G
APOSOR1336-1GEA 165 GBP
APOSOR1336-1G APOSOR1336-250MG APOSOR1336-5G
3-Amino-4-chlorophenylboronic acid pinacol ester 98%
3-Amino-4-chlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₇BClNO₂
MW: 253,54 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD06659926
CAS Number: 850567-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR