(±)-3-Amino-3-phenylpropionic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
DL-β-Phenylalanine, 3-Amino-3-phenylpropionic acid, H-DL-β-Phe-OH, β-Aminohydrocinnamic acid
OR13352-100G
OR13352-25G
OR13352-5G
APOSOR13352-100GEA
70.9
GBP
APOSOR13352-100G
APOSOR13352-25G
APOSOR13352-5G
(±)-3-Amino-3-phenylpropionic acid
(±)-3-Amino-3-phenylpropionic acid
Formula:
C₉H₁₁NO₂ MW: 165,19 g/mol Melting Pt: 222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008064 CAS Number: 614-19-7 EINECS: 210-371-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...