Methyl cyanoacetate 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 2-cyanoacetate
OR13190-100G
OR13190-1KG
OR13190-500G
APOSOR13190-100GEA
52.5
GBP
APOSOR13190-100G
APOSOR13190-1KG
APOSOR13190-500G
Methyl cyanoacetate 99%
Methyl cyanoacetate
Formula:
C₄H₅NO₂ MW: 99,09 g/mol Boiling Pt: 204…207 °C Melting Pt: –13 °C Density: 1,123 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001939 CAS Number: 105-34-0 EINECS: 203-288-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...