3-Iodopyridine

Supplier: Apollo Scientific
Warning

OR13049-100G OR13049-25G
APOSOR13049-100GEA 86.4 GBP
APOSOR13049-100G APOSOR13049-25G
3-Iodopyridine
3-Iodopyridine
Formula: C₅H₄IN
Melting Pt: 53…56 °C
Flash Pt: 106,7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023553
CAS Number: 1120-90-7
EINECS: 214-322-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR