Methyl-2,4-dichloropyrimidine-5-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2,4-dichloropyrimidine-5-carboxylate

OR12636-1G OR12636-5G
APOSOR12636-1GEA 61.7 GBP
APOSOR12636-1G APOSOR12636-5G
Methyl-2,4-dichloropyrimidine-5-carboxylate
Methyl-2,4-dichloropyrimidine-5-carboxylate
Formula: C₆H₄Cl₂N₂O₂
MW: 207,0142 g/mol
Boiling Pt: 141…143 °C/23mmHg
Melting Pt: 30…34 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09910342
CAS Number: 3177-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR