2-(Pyrrolidin-3-yl)pyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(Pyridin-2-yl)pyrrolidine
OR12553-1G
OR12553-5G
APOSOR12553-1GEA
223
GBP
APOSOR12553-1G
APOSOR12553-5G
2-(Pyrrolidin-3-yl)pyridine
2-(Pyrrolidin-3-yl)pyridine
Formula:
C₉H₁₂N₂ MW: 148.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01862539 CAS Number: 150281-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...