5-Methylisoxazole-4-carbonyl chloride 95%
Supplier: Apollo Scientific
|
Danger
|
OR12344-1G
OR12344-250MG
APOSOR12344-1GEA
217
GBP
APOSOR12344-1G
APOSOR12344-250MG
5-Methylisoxazole-4-carbonyl chloride 95%
5-Methylisoxazole-4-carbonyl chloride
Formula:
C₅H₄ClNO₂ MW: 145,55 g/mol Boiling Pt: 78…79 °C/14mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD03411599 CAS Number: 67305-24-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...