4-Morpholinyl(2-piperidinyl)methanone hydrochloride

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Piperidin-2-ylcarbonyl)morpholine hydrochloride

OR12325-1G OR12325-250MG
APOSOR12325-1GEA 191 GBP
APOSOR12325-1G APOSOR12325-250MG
4-Morpholinyl(2-piperidinyl)methanone hydrochloride
4-Morpholinyl(2-piperidinyl)methanone hydrochloride
Formula: C₁₀H₁₉ClN₂O₂
MW: 234,73 g/mol
Melting Pt: 291…293 °C
Storage Temperature: Ambient
MDL Number: MFCD05865124
CAS Number: 690634-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR