2-(Chloromethyl)-4-nitrobenzoxazole

Supplier: Apollo Scientific

OR12179-1G OR12179-10G OR12179-5G
APOSOR12179-1GEA 0 GBP
APOSOR12179-1G APOSOR12179-10G APOSOR12179-5G
2-(Chloromethyl)-4-nitrobenzoxazole
2-(Chloromethyl)-4-nitrobenzoxazole
Formula: C₈H₅ClN₂O₃
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD09027181
CAS Number: 143708-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR