5-Amino-6-bromo-2-pyridinecarbonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Amino-6-bromopicolinonitrile
OR12165-1G
OR12165-250MG
APOSOR12165-1GEA
491
GBP
APOSOR12165-1G
APOSOR12165-250MG
5-Amino-6-bromo-2-pyridinecarbonitrile
5-Amino-6-bromo-2-pyridinecarbonitrile
Formula:
C₆H₄BrN₃ MW: 198,03 g/mol Melting Pt: 172…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD09027188 CAS Number: 849353-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...