3-(4H-1,2,4-Triazol-4-yl)-1-propanol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(3-Hydroxyprop-1-yl)-4H-1,2,4-triazole
OR12163-1G
OR12163-250MG
OR12163-5G
APOSOR12163-1GEA
263
GBP
APOSOR12163-1G
APOSOR12163-250MG
APOSOR12163-5G
3-(4H-1,2,4-Triazol-4-yl)-1-propanol
3-(4H-1,2,4-Triazol-4-yl)-1-propanol
Formula:
C₅H₉N₃O MW: 127,15 g/mol Melting Pt: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840804 CAS Number: 27106-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...