5-Nitro-2-piperidinobenzenecarbonitrile
Supplier: Apollo Scientific
Synonyms:
5-Nitro-2-piperidin-1-ylbenzonitrile
OR12156-1G
OR12156-5G
APOSOR12156-1GEA
85.2
GBP
APOSOR12156-1G
APOSOR12156-5G
5-Nitro-2-piperidinobenzenecarbonitrile
5-Nitro-2-piperidinobenzenecarbonitrile
Formula:
C₁₂H₁₃N₃O₂ MW: 231,26 g/mol Melting Pt: 56…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD05365582 CAS Number: 32188-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...