1-(2-Bromoethoxy)-3-methoxybenzene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(2-Bromoethoxy)anisole
OR12039-1G
OR12039-25G
OR12039-5G
APOSOR12039-1GEA
13.2
GBP
APOSOR12039-1G
APOSOR12039-25G
APOSOR12039-5G
1-(2-Bromoethoxy)-3-methoxybenzene
1-(2-Bromoethoxy)-3-methoxybenzene
Formula:
C₉H₁₁O₂Br MW: 231,0864 g/mol Boiling Pt: 84 °C/0,023mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00980056 CAS Number: 3245-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...