1-(Benzyloxy)-4-(2-bromoethoxy)benzene

Supplier: Apollo Scientific
Warning

OR12023-1G OR12023-5G OR12023-10G
APOSOR12023-1GEA 139 GBP
APOSOR12023-1G APOSOR12023-5G APOSOR12023-10G
1-(Benzyloxy)-4-(2-bromoethoxy)benzene
1-(Benzyloxy)-4-(2-bromoethoxy)benzene
Formula: C₁₅H₁₅BrO₂
MW: 307,19 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD08689757
CAS Number: 3351-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR