2-Amino-1-morpholino-1-ethanone hydrochloride

Supplier: Apollo Scientific

Synonyms: 4-(Aminoacetyl)morpholine hydrochloride, 2-Amino-1-morpholinoethanone hydrochloride

OR12016-1G OR12016-5G OR12016-10G
APOSOR12016-1GEA 108 GBP
APOSOR12016-1G APOSOR12016-5G APOSOR12016-10G
2-Amino-1-morpholino-1-ethanone hydrochloride
2-Amino-1-morpholino-1-ethanone hydrochloride
Formula: C₆H₁₃ClN₂O₂
MW: 180,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366740
CAS Number: 24152-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR