5-Methylisophthalic acid
Supplier: Apollo Scientific
|
Warning
|
OR12006-100G
OR12006-25G
OR12006-5G
APOSOR12006-100GEA
60.2
GBP
APOSOR12006-100G
APOSOR12006-25G
APOSOR12006-5G
5-Methylisophthalic acid
5-Methylisophthalic acid
Formula:
C₉H₈O₄ Melting Pt: 299…303 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013986 CAS Number: 499-49-0 EINECS: 207-881-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...