2-Chloro-5-methoxyphenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-5-methoxyphenylboronic Acid, 2-Chloro-5-methoxybenzeneboronic acid

OR1195-5G OR1195-25G
APOSOR1195-5GEA 25 GBP
APOSOR1195-5G APOSOR1195-25G
2-Chloro-5-methoxyphenylboronic acid
2-Chloro-5-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MW: 186,40 g/mol
Melting Pt: 106…112 °C
Storage Temperature: Ambient
MDL Number: MFCD06659858
CAS Number: 89694-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR