5-Bromo-N-isopropylpicolinamide 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-N-isopropylpyridine-2-carboxamide

OR11920-1G OR11920-5G
APOSOR11920-1GEA 129 GBP
APOSOR11920-1G APOSOR11920-5G
5-Bromo-N-isopropylpicolinamide 98%
5-Bromo-N-isopropylpicolinamide
Formula: C₉H₁₁BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09801058
CAS Number: 845305-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR