3-Bromo-4-hydroxybenzaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
OR1184-25G
OR1184-5G
OR1184-100G
APOSOR1184-25GEA
47.1
GBP
APOSOR1184-25G
APOSOR1184-5G
APOSOR1184-100G
3-Bromo-4-hydroxybenzaldehyde 97%
3-Bromo-4-hydroxybenzaldehyde
Formula:
C₇H₅BrO₂ MW: 201.02 g/mol Melting Pt: 125…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017348 CAS Number: 2973-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...