2-Cyano-3-methylpyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Methylpyridine-2-carbonitrile, 3-Methylpicolinonitrile, 2-Cyano-β-picoline, 2-Cyano-3-picoline
OR11740-100G
OR11740-25G
APOSOR11740-100GEA
60.3
GBP
APOSOR11740-100G
APOSOR11740-25G
2-Cyano-3-methylpyridine
2-Cyano-3-methylpyridine
Formula:
C₇H₆N₂ MW: 118,14 g/mol Boiling Pt: 141 °C/38mmHg Melting Pt: 83…87 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190585 CAS Number: 20970-75-6 EINECS: 244-131-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...