[3-(Bromomethyl)oxetan-3-yl]methanol
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(3-(Bromomethyl)oxetan-3-yl)methanol
OR11736-1G
OR11736-5G
APOSOR11736-1GEA
0
GBP
APOSOR11736-1G
APOSOR11736-5G
[3-(Bromomethyl)oxetan-3-yl]methanol
[3-(Bromomethyl)oxetan-3-yl]methanol
Formula:
C₅H₉BrO₂ MW: 181,04 g/mol Boiling Pt: 242 °C Density: 1,598 g/cm³ (25 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD09800598 CAS Number: 22633-44-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...