2-Bromoisonicotinonitrile 95%
Supplier: Apollo Scientific
|
Warning
|
OR11615-1G
OR11615-5G
OR11615-25G
APOSOR11615-1GEA
22.3
GBP
APOSOR11615-1G
APOSOR11615-5G
APOSOR11615-25G
2-Bromoisonicotinonitrile 95%
2-Bromoisonicotinonitrile
Formula:
C₆H₃BrN₂ Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD07367879 CAS Number: 10386-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...