2-Bromotoluene-4-sulphonamide 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-4-methylbenzenesulphonamide
OR11601-1G
OR11601-5G
APOSOR11601-1GEA
156
GBP
APOSOR11601-1G
APOSOR11601-5G
2-Bromotoluene-4-sulphonamide 98%
2-Bromotoluene-4-sulphonamide
Formula:
C₇H₈BrNO₂S MW: 250,1129 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09801033 CAS Number: 210824-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...