4-Bromo-N-tert-butylbenzamide 98%

Supplier: Apollo Scientific

Synonyms: 4-Bromo-N-(tert-butyl)benzamide

OR11562-1G OR11562-5G
APOSOR11562-1GEA 64.3 GBP
APOSOR11562-1G APOSOR11562-5G
4-Bromo-N-tert-butylbenzamide 98%
4-Bromo-N-tert-butylbenzamide
Formula: C₁₁H₁₄BrNO
MW: 256,1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462539
CAS Number: 42498-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR