Ethyl-7-nitroindole-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 7-nitro-1H-indole-2-carboxylate
OR1145-1G
OR1145-5G
OR1145-10G
APOSOR1145-1GEA
32.8
GBP
APOSOR1145-1G
APOSOR1145-5G
APOSOR1145-10G
Ethyl-7-nitroindole-2-carboxylate
Ethyl-7-nitroindole-2-carboxylate
Formula:
C₁₁H₁₀N₂O₄ MW: 234,211 g/mol Melting Pt: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216475 CAS Number: 6960-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...