1-(2-Bromoethoxy)-3-nitrobenzene 97%

Supplier: Apollo Scientific
Warning

OR11413-1G OR11413-25G OR11413-5G
APOSOR11413-1GEA 13.2 GBP
APOSOR11413-1G APOSOR11413-25G APOSOR11413-5G
1-(2-Bromoethoxy)-3-nitrobenzene 97%
1-(2-Bromoethoxy)-3-nitrobenzene
Formula: C₈H₈BrNO₃
MW: 246,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783648
CAS Number: 13831-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR