4-Methylthiazole-5-carboxylic acid 95%
Supplier: Apollo Scientific
|
Warning
|
OR11384-10G
OR11384-1G
OR11384-100G
APOSOR11384-10GEA
23.6
GBP
APOSOR11384-10G
APOSOR11384-1G
APOSOR11384-100G
4-Methylthiazole-5-carboxylic acid 95%
4-Methylthiazole-5-carboxylic acid
Formula:
C₅H₅NO₂S MW: 143.17 g/mol Melting Pt: 287 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00626872 CAS Number: 20485-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...