3,5-Dimethoxybenzyl alcohol 98%

Supplier: Apollo Scientific

Synonyms: (3,5-Dimethoxyphenyl)methanol

OR11308-100G OR11308-10G OR11308-25G
APOSOR11308-100GEA 91.8 GBP
APOSOR11308-100G APOSOR11308-10G APOSOR11308-25G
3,5-Dimethoxybenzyl alcohol 98%
3,5-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
MW: 168,19 g/mol
Boiling Pt: 183…186 °C/20mmHg
Melting Pt: 46…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00004641
CAS Number: 705-76-0
EINECS: 211-888-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR