4-Butylphenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-n-Butylbenzeneboronic acid

OR11251-25G OR11251-5G
APOSOR11251-25GEA 84 GBP
APOSOR11251-25G APOSOR11251-5G
4-Butylphenylboronic acid 98%
4-Butylphenylboronic acid
Formula: C₁₀H₁₅BO₂
MW: 178,04 g/mol
Melting Pt: 91…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093926
CAS Number: 145240-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR