3'-Nitroacetophenone 98%

Supplier: Apollo Scientific
Warning

Synonyms: m-Nitroacetophenone

OR11221-1KG OR11221-250G
APOSOR11221-1KGEA 0 GBP
APOSOR11221-1KG APOSOR11221-250G
3'-Nitroacetophenone 98%
3'-Nitroacetophenone
Formula: C₈H₇NO₃
MW: 165,15 g/mol
Boiling Pt: 202 °C
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00007259
CAS Number: 121-89-1
EINECS: 204-504-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR