Diethyl-3-oxoglutarate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate
OR11166-100G
OR11166-25G
OR11166-5G
APOSOR11166-100GEA
108
GBP
APOSOR11166-100G
APOSOR11166-25G
APOSOR11166-5G
Diethyl-3-oxoglutarate 97%
Diethyl 3-oxoglutarate
Formula:
C₉H₁₄O₅ MW: 202,21 g/mol Boiling Pt: 250 °C Density: 1,112 g/cm³ (25 °C) Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009200 CAS Number: 105-50-0 EINECS: 203-302-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...