2-Bromo-1-methoxynaphthalene
Supplier: Apollo Scientific
|
Warning
|
OR111505-100MG
OR111505-1G
OR111505-250MG
APOSOR111505-100MGEA
52.5
GBP
APOSOR111505-100MG
APOSOR111505-1G
APOSOR111505-250MG
2-Bromo-1-methoxynaphthalene
2-Bromo-1-methoxynaphthalene
Formula:
C₁₁H₉BrO Storage Temperature: Ambient |
MDL Number:
MFCD17012354 CAS Number: 62012-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...